COM448612197 Combi-Blocks, Inc.

4-Bromo-2-methyl-6-nitroanisole

  • MDL number MFCD22205732
  • Cas Number : 1375069-27-4
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2953
Availability details for
Quantity
  • Availability
  • Add to Cart