COM448612198 Combi-Blocks, Inc.

1-Methyl-4-(3-nitrophenyl)piperazine

  • MDL number MFCD08435858
  • Cas Number : 148546-97-8
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2954
Availability details for
Quantity
  • Availability
  • Add to Cart