COM448612199 Combi-Blocks, Inc.

1,2-Dibromo-3-nitrobenzene

  • MDL number MFCD00049051
  • Cas Number : 26429-41-4
  • Molecular Weight : 280.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2955
Availability details for
Quantity
  • Availability
  • Add to Cart