COM448612210 Combi-Blocks, Inc.

4-Bromo-N-cyclohexyl-5-ethoxy-2-nitroaniline

  • MDL number MFCD21609504
  • Cas Number : 1365272-39-4
  • Molecular Weight : 343.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2976

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2976
Availability details for
Quantity
  • Availability
  • Add to Cart