COM448612233 Combi-Blocks, Inc.

4-[(Azepan-1-yl)carbonyl]benzylamine

  • MDL number MFCD08444569
  • Cas Number : 923120-30-3
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3007
Availability details for
Quantity
  • Availability
  • Add to Cart