COM448612234 Combi-Blocks, Inc.

4-(Azetidinocarbonyl)benzylamine

  • MDL number MFCD08444577
  • Cas Number : 923183-92-0
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3008
Availability details for
Quantity
  • Availability
  • Add to Cart