COM448612237 Combi-Blocks, Inc.

N-Cyclohexyl 4-(aminomethyl)benzamide

  • MDL number MFCD09045342
  • Cas Number : 172735-66-9
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3011
Availability details for
Quantity
  • Availability
  • Add to Cart