COM448612241 Combi-Blocks, Inc.

N-Cyclopropyl 4-(aminomethyl)benzamide

  • MDL number MFCD09043807
  • Cas Number : 789489-39-0
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3015

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3015
Availability details for
Quantity
  • Availability
  • Add to Cart