COM448612249 Combi-Blocks, Inc.

1,3-Dimethyl 2-(2-amino-4-cyanophenyl)propanedioate

  • MDL number MFCD22192349
  • Cas Number : 1373232-54-2
  • Molecular Weight : 248.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3023
Availability details for
Quantity
  • Availability
  • Add to Cart