COM448612257 Combi-Blocks, Inc.

3-Amino-5-bromo-N,N-dimethylbenzamide

  • MDL number MFCD22192353
  • Cas Number : 1369784-97-3
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3031
Availability details for
Quantity
  • Availability
  • Add to Cart