COM448612259 Combi-Blocks, Inc.

3-Amino-5-bromobenzamide

  • MDL number MFCD18389345
  • Cas Number : 1261609-26-0
  • Molecular Weight : 215.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3033
Availability details for
Quantity
  • Availability
  • Add to Cart