COM448612276 Combi-Blocks, Inc.

3-Amino-5-bromo-N-methylbenzamide

  • MDL number MFCD22205741
  • Cas Number : 1375069-14-9
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3053
Availability details for
Quantity
  • Availability
  • Add to Cart