COM448612281 Combi-Blocks, Inc.

3-(Benzyloxy)-4-chloroaniline

  • MDL number MFCD19625037
  • Cas Number : 76464-57-8
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3058

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3058
Availability details for
Quantity
  • Availability
  • Add to Cart