COM448612294 Combi-Blocks, Inc.

3-Bromo-4-hydroxy-5-nitrobenzoic acid

  • MDL number MFCD14708201
  • Cas Number : 67175-27-3
  • Molecular Weight : 262.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3107
Availability details for
Quantity
  • Availability
  • Add to Cart