COM448612309 Combi-Blocks, Inc.

N-Ethyl-3-nitrobenzamide

  • MDL number MFCD01211875
  • Cas Number : 26819-11-4
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3125

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3125
Availability details for
Quantity
  • Availability
  • Add to Cart