COM448612318 Combi-Blocks, Inc.

methyl 2-methoxy-3-nitrobenzoate

  • MDL number MFCD11045625
  • Cas Number : 90564-26-4
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3134
Availability details for
Quantity
  • Availability
  • Add to Cart