COM448612323 Combi-Blocks, Inc.

5-Bromo-2-ethoxy-3-methylaniline

  • MDL number MFCD22375005
  • Cas Number : 1381944-55-3
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3139

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3139
Availability details for
Quantity
  • Availability
  • Add to Cart