COM448612329 Combi-Blocks, Inc.

1-Bromo-2-chloro-5-(cyclopropylmethoxy)-4-nitrobenzene

  • MDL number MFCD22421476
  • Cas Number : 1400644-63-4
  • Molecular Weight : 306.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3148
Availability details for
Quantity
  • Availability
  • Add to Cart