COM448612333 Combi-Blocks, Inc.

5-Bromo-4-chloro-2-nitroanisole

  • MDL number MFCD22383655
  • Cas Number : 1360616-77-8
  • Molecular Weight : 266.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3152
Availability details for
Quantity
  • Availability
  • Add to Cart