COM448612342 Combi-Blocks, Inc.

1-Methyl-4-(2-methyl-4-nitrophenyl)piperazine

  • MDL number MFCD11039654
  • Cas Number : 681004-49-9
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3162

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3162
Availability details for
Quantity
  • Availability
  • Add to Cart