COM448612353 Combi-Blocks, Inc.

1-(3-amino-2-methylphenyl)pyrrolidin-2-one

  • MDL number MFCD09044451
  • Cas Number : 69131-43-7
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3175
Availability details for
Quantity
  • Availability
  • Add to Cart