COM448612354 Combi-Blocks, Inc.

2-(Benzyloxy)-1-bromo-5-fluoro-3-nitrobenzene

  • MDL number MFCD22383657
  • Cas Number : 1393441-89-8
  • Molecular Weight : 326.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3176

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3176
Availability details for
Quantity
  • Availability
  • Add to Cart