COM448612366 Combi-Blocks, Inc.

(5-Bromo-4-chloro-2-nitrophenyl)acetic acid

  • MDL number MFCD22421480
  • Cas Number : 1400644-94-1
  • Molecular Weight : 294.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3198

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3198
Availability details for
Quantity
  • Availability
  • Add to Cart