COM448612367 Combi-Blocks, Inc.

4-[(4-Fluoro-3-nitrobenzene)sulfonyl]morpholine

  • MDL number MFCD21366492
  • Cas Number : 1338346-30-7
  • Molecular Weight : 290.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3200
Availability details for
Quantity
  • Availability
  • Add to Cart