COM448612390 Combi-Blocks, Inc.

2,4-Dichloro-3-methyl-1,5-dinitrobenzene

  • MDL number MFCD08582840
  • Cas Number : 51676-76-7
  • Molecular Weight : 251.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3236
Availability details for
Quantity
  • Availability
  • Add to Cart