COM448612401 Combi-Blocks, Inc.

2-Amino-5-bromo-4-chlorobenzamide

  • MDL number MFCD20037897
  • Cas Number : 1379338-88-1
  • Molecular Weight : 249.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3251

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3251
Availability details for
Quantity
  • Availability
  • Add to Cart