COM448612402 Combi-Blocks, Inc.

5-Bromo-4-chloro-2-nitrobenzamide

  • MDL number MFCD28127174
  • Cas Number : 1820608-87-4
  • Molecular Weight : 279.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3254

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3254
Availability details for
Quantity
  • Availability
  • Add to Cart