COM448612415 Combi-Blocks, Inc.

3-(Morpholin-4-yl)-4-nitrobenzoic acid

  • MDL number MFCD12031201
  • Cas Number : 1134335-58-2
  • Molecular Weight : 252.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3268

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3268
Availability details for
Quantity
  • Availability
  • Add to Cart