COM448612426 Combi-Blocks, Inc.

4-(4-Bromo-2-methoxyphenoxy)-1-methylpiperidine

  • MDL number MFCD22549306
  • Cas Number : 1403483-71-5
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3290

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3290
Availability details for
Quantity
  • Availability
  • Add to Cart