COM448612427 Combi-Blocks, Inc.

N,N-Dimethyl 5-bromo-2-nitrobenzylamine

  • MDL number MFCD22421487
  • Cas Number : 1400644-49-6
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3291
Availability details for
Quantity
  • Availability
  • Add to Cart