COM448612428 Combi-Blocks, Inc.

4-Nitro-3-(phenylamino)benzoic acid

  • MDL number MFCD22549307
  • Cas Number : 159190-73-5
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3295

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3295
Availability details for
Quantity
  • Availability
  • Add to Cart