COM448612431 Combi-Blocks, Inc.

2-Fluoro-4-(1-methylhydrazin-1-yl)benzonitrile

  • MDL number MFCD22549309
  • Cas Number : 1357626-46-0
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3302
Availability details for
Quantity
  • Availability
  • Add to Cart