COM448612432 Combi-Blocks, Inc.

Ethyl 2-[(4-bromo-2-nitrophenyl)amino]acetate

  • MDL number MFCD12728171
  • Cas Number : 1179891-92-9
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3303

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3303
Availability details for
Quantity
  • Availability
  • Add to Cart