COM448612433 Combi-Blocks, Inc.

N-BOC-3-Methoxy-4-nitroaniline

  • MDL number MFCD22549310
  • Cas Number : 877671-39-1
  • Molecular Weight : 268.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3304
Availability details for
Quantity
  • Availability
  • Add to Cart