COM448612438 Combi-Blocks, Inc.

Methyl 3-(butylamino)-4-nitrobenzoate

  • MDL number MFCD22689082
  • Cas Number : 597562-38-4
  • Molecular Weight : 252.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3311
Availability details for
Quantity
  • Availability
  • Add to Cart