COM448612457 Combi-Blocks, Inc.

4-Bromo-2-nitrobenzene-1,3-diamine

  • MDL number MFCD22689085
  • Cas Number : 1420800-18-5
  • Molecular Weight : 232.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3336

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3336
Availability details for
Quantity
  • Availability
  • Add to Cart