COM448612471 Combi-Blocks, Inc.

N-Pivaloyl 4-bromo-3-fluoro-2-nitroaniline

  • MDL number MFCD22689087
  • Cas Number : 1420800-43-6
  • Molecular Weight : 319.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3351
Availability details for
Quantity
  • Availability
  • Add to Cart