COM448612472 Combi-Blocks, Inc.

2-N-Butylpyridine-2,3-diamine

  • MDL number MFCD11101082
  • Cas Number : 53929-75-2
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3352
Availability details for
Quantity
  • Availability
  • Add to Cart