COM448612479 Combi-Blocks, Inc.

3-(N,N-Dimethylamino)-4-methanesulfonylaniline

  • MDL number MFCD22689088
  • Cas Number : 1420800-40-3
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3362
Availability details for
Quantity
  • Availability
  • Add to Cart