COM448612487 Combi-Blocks, Inc.

N-(4-Bromo-2-nitrophenyl)-2-chloroacetamide

  • MDL number MFCD07468929
  • Cas Number : 854583-85-0
  • Molecular Weight : 293.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3377
Availability details for
Quantity
  • Availability
  • Add to Cart