COM448612497 Combi-Blocks, Inc.

2,5-Dimethyl 3-nitrothiophene-2,5-dicarboxylate

  • MDL number MFCD22381980
  • Cas Number : 858840-42-3
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3400

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3400
Availability details for
Quantity
  • Availability
  • Add to Cart