COM448612510 Combi-Blocks, Inc.

2-(4-Cyclopropylpiperazine-1-sulfonyl)aniline

  • MDL number MFCD16369332
  • Cas Number : 1290734-17-6
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3420
Availability details for
Quantity
  • Availability
  • Add to Cart