COM448612511 Combi-Blocks, Inc.

4-Ethanesulfonyl-3-morpholinonitrobenzene

  • MDL number MFCD25953890
  • Cas Number : 1820641-46-0
  • Molecular Weight : 300.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3421
Availability details for
Quantity
  • Availability
  • Add to Cart