COM448612526 Combi-Blocks, Inc.

3-Bromo-5-fluoroaniline HCl

  • MDL number MFCD09800744
  • Cas Number : 134168-98-2
  • Molecular Weight : 226.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3437
Availability details for
Quantity
  • Availability
  • Add to Cart