COM448612529 Combi-Blocks, Inc.

2-(2-Aminoethoxy)-1,3-dichlorobenzene

  • MDL number MFCD06212826
  • Cas Number : 17944-28-4
  • Molecular Weight : 206.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3446

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3446
Availability details for
Quantity
  • Availability
  • Add to Cart