COM448612546 Combi-Blocks, Inc.

4-Bromo-2-nitro-N-phenylaniline

  • MDL number MFCD18579563
  • Cas Number : 16588-25-3
  • Molecular Weight : 293.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3471
Availability details for
Quantity
  • Availability
  • Add to Cart