COM448612568 Combi-Blocks, Inc.

4-Bromo-5-nitro-1,2-methylenedioxybenzene

  • MDL number MFCD00156317
  • Cas Number : 7748-58-5
  • Molecular Weight : 246.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3502
Availability details for
Quantity
  • Availability
  • Add to Cart