COM448612574 Combi-Blocks, Inc.

4-Methanesulfonyl-2-methylaniline

  • MDL number MFCD09909541
  • Cas Number : 252562-00-8
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3519

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3519
Availability details for
Quantity
  • Availability
  • Add to Cart