COM448612575 Combi-Blocks, Inc.

1-Bromo-3-fluoro-2-methyl-4-nitrobenzene

  • MDL number MFCD27935488
  • Cas Number : 1806979-60-1
  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3521
Availability details for
Quantity
  • Availability
  • Add to Cart