COM448612580 Combi-Blocks, Inc.

2-(5-Bromo-3-fluoro-2-nitrophenyl)acetonitrile

  • MDL number MFCD27981294
  • Cas Number : 1807211-69-3
  • Molecular Weight : 259.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3529
Availability details for
Quantity
  • Availability
  • Add to Cart